3-Bromo-5-nitrotoluene

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Bromo-3-methyl-5-nitrobenzene

ACTIAS40326G5EA 0 CHF
ACTIAS40326G5
3-Bromo-5-nitrotoluene
3-Bromo-5-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD06659815
CAS Number: 52488-28-5
EINECS: 257-955-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR