2-Amino-3-bromo-6-methylpyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS7219G5EA 0 CHF
ACTIAS7219G1 ACTIAS7219G25 ACTIAS7219G5
2-Amino-3-bromo-6-methylpyridine
2-Amino-3-bromo-6-methylpyridine
Formula: C₆H₇BrN₂
MDL Number: MFCD03095215
CAS Number: 126325-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR