2-Amino-3-brompyridin
Lieferant: ACTIVATE SCIENTIFIC GMBH
|
Achtung
|
ACTIAS40148G1EA
0
CHF
ACTIAS40148G1
ACTIAS40148G5
2-Amino-3-brompyridin
2-Amino-3-brompyridin
Formel:
C₅H₅BrN₂ MW: 173,01 g/mol |
MDL Number:
MFCD03095187 CAS-Nummer: 13534-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...