4-Bromo-2,6-xylidine

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Bromo-2,6-dimethylaniline

ACTIAS40392G100EA 0 CHF
ACTIAS40392G100
4-Bromo-2,6-xylidine
4-Bromo-2,6-xylidine
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 120 °C (0.2 mmHg)
Melting Pt: 50 °C
MDL Number: MFCD00007826
CAS Number: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR