3-Bromobenzylamine

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: (3-Bromophenyl)methanamine

ACTIAS33326G5EA 0 CHF
ACTIAS33326G10 ACTIAS33326G5
3-Bromobenzylamine
3-Bromobenzylamine
Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD01026119
CAS Number: 10269-01-9
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR