4-Brom-3-nitrobenzotrifluorid

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Brom-2-nitro-4-(trifluormethyl)benzol4-Brom-3-nitrobenzotrifluorid

ACTIAS40411G100EA 0 CHF
ACTIAS40411G100
4-Brom-3-nitrobenzotrifluorid
4-Brom-3-nitrobenzotrifluorid
Formel: C₇H₃BrF₃NO₂
MW: 270,01 g/mol
Siedepunkt: 102…105 °C (10 mmHg)
Dichte: 1,76 g/cm³ (20 °C)
MDL Number: MFCD00014685
CAS-Nummer: 349-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR