5-Brom-1-indanon

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS20012G5EA 0 CHF
ACTIAS20012G50 ACTIAS20012G5 ACTIAS20012G10
5-Brom-1-indanon
5-Brom-1-indanon
Formel: C₉H₇BrO
MW: 211,06 g/mol
Siedepunkt: 280 °C (1013 hPa)
Melting Pt: 126…127 °C
MDL Number: MFCD00082718
CAS-Nummer: 34598-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR