3-Brombenzophenon

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS23499G25EA 0 CHF
ACTIAS23499G25 ACTIAS23499G5
3-Brombenzophenon
3-Brombenzophenon
Formel: C₁₃H₉BrO
MW: 261,12 g/mol
Siedepunkt: 355 °C (1013 hPa)
Melting Pt: 74,5…77.5 °C
Dichte: 1,42 g/cm³ (20 °C)
MDL Number: MFCD00672015
CAS-Nummer: 1016-77-9
EINECS: 213-808-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR