1-Brom-2-chlor-4-fluorbenzol

Lieferant: ACTIVATE SCIENTIFIC GMBH
Achtung

Synonyms: 4-Brom-3-chlor-1-fluorbenzol

ACTIAS40073G100EA 0 CHF
ACTIAS40073G25 ACTIAS40073G100
1-Brom-2-chlor-4-fluorbenzol
1-Bromo-2-chloro-4-fluorobenzene
Formel: C₆H₃BrClF
MW: 209,44 g/mol
MDL Number: MFCD00142587
CAS-Nummer: 110407-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR