3-Brombiphenyl (PBB Nr. 2)

Lieferant: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1-Brom-3-phenylbenzol

ACTIAS51071G5EA 0 CHF
ACTIAS51071G5 ACTIAS51071G25
3-Brombiphenyl (PBB Nr. 2)
3-Brombiphenyl (PBB Nr. 2)
Formel: C₁₂H₉Br
MW: 233,11 g/mol
Siedepunkt: 299 °C (1013 hPa)
Melting Pt: 9 °C
Dichte: 1,41 g/cm³ (20 °C)
MDL Number: MFCD00000082
CAS-Nummer: 2113-57-7
EINECS: 218-304-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR