3,5-Dimethyl-1H-pyrazole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 3,5-Dimethylpyrazol3,5-Dimethylpyrazole3,5-Dimethyl-1H-pyrazole

ACTIAS40293G100EA 101 CHF
ACTIAS40293G100
3,5-Dimethyl-1H-pyrazole
3,5-Dimethyl-1H-pyrazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 218 °C (1013 hPa)
Density: 0,8839 g/cm³ (16 °C)
MDL Number: MFCD00005243
CAS Number: 67-51-6
EINECS: 200-657-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR