Dibenzofuran

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Dibenzo[b,d]furan

ACTIAS24276G100EA 199 CHF
ACTIAS24276G100
Dibenzofuran
Dibenzofuran
Formula: C₁₂H₈O
MW: 168.19 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 79…82 °C
Density: 0.966 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004968
CAS Number: 132-64-9
EINECS: 205-071-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR