1-Vinylnaphthalene

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS23928G5EA 0 CHF
ACTIAS23928G5 ACTIAS23928G25
1-Vinylnaphthalene
1-Vinylnaphthalene
Formula: C₁₂H₁₀
MW: 154.21 g/mol
Boiling Pt: 271 °C (1013 hPa)
Melting Pt: 21 °C
Density: 1.032 g/cm³ (20 °C)
Storage Temperature: Freezer
MDL Number: MFCD00075766
CAS Number: 826-74-4
EINECS: 212-560-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR