Dipotassium hydrogen orthophosphate trihydrate
Supplier: VWR Chemicals
Synonyms: Potassium hydrogen phosphate trihydratePotassium phosphate dibasic trihydratedi-Potassium hydrogen orthophosphate trihydratedi-Potassium hydrogen phosphate trihydrate
K967-1KGEA 278 CHF
K967-1KG
Dipotassium hydrogen orthophosphate trihydrate
di-Potassium hydrogen orthophosphate trihydrate
Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
Boiling Pt: ∼340 °C (1013 hPa)
Density: 2.504 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5