(R)-(-)-1,2-Propandiol
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
(R)-(-)-1,2-Dihydroxypropan
, (R)-(-)-1,2-Propylenglycol
, (R)-(-)-1,2-Propylenglykol
ACTIAS20999G25EA
0
CHF
ACTIAS20999G25
ACTIAS20999G100
(R)-(-)-1,2-Propandiol
(R)-(-)-1,2-Propandiol
Formel:
C₃H₈O₂ MW: 76,1 g/mol |
MDL Number:
MFCD00066248 CAS-Nummer: 4254-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...