Hypoxanthine
Supplier: ACTIVATE SCIENTIFIC GMBH
Synonyms:
1H-Purin-6-one
, Purin-6(1H)-one
, 6-Purinone
ACTIAS23792G100EA
0
CHF
ACTIAS23792G100
Hypoxanthine
Hypoxanthine
Formula:
C₅H₄N₄O MW: 136.11 g/mol Melting Pt: > 310 °C |
MDL Number:
MFCD00005725 CAS Number: 68-94-0 EINECS: 200-697-3 Merck Index: 13,04895 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...