4-Indanol

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS32652G5EA 0 CHF
ACTIAS32652G5
4-Indanol
4-Indanol
Formel: C₉H₁₀O
MW: 134,18 g/mol
MDL Number: MFCD00053431
CAS-Nummer: 1641-41-4
EINECS: 216-691-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR