2,3-Dimethoxypyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS4011G5EA 0 CHF
ACTIAS4011G25 ACTIAS4011G5
2,3-Dimethoxypyridine
2,3-Dimethoxypyridine
Formula: C₇H₉NO₂
MW: 139.15 g/mol
MDL Number: MFCD03095043
CAS Number: 52605-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR