(R)-(-)-1-Methoxy-2-propanol

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: (R)-1-Methoxy-2-propanol(R)-(-)-Propylene glycol 1-Monomethyl ether

ACTIAS31704G5EA 0 CHF
ACTIAS31704G10 ACTIAS31704G5
(R)-(-)-1-Methoxy-2-propanol
(R)-(-)-1-Methoxy-2-propanol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01632587
CAS Number: 4984-22-9
UN: 3092
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR