Methanesulphonic anhydride
Supplier: ACTIVATE SCIENTIFIC GMBH
|
Danger
|
ACTIAS24108G100EA
0
CHF
ACTIAS24108G500
ACTIAS24108G100
Methanesulphonic anhydride
Methanesulphonic anhydride
Formula:
C₂H₆O₅S₂ MW: 174.2 g/mol Melting Pt: 63…66 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007556 CAS Number: 7143-01-3 EINECS: 230-442-1 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...