4-Nitroindole

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS21444G5EA 34 CHF
ACTIAS21444G5 ACTIAS21444G1
4-Nitroindole
4-Nitroindole
Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 206…209 °C
MDL Number: MFCD00010056
CAS Number: 4769-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR