2-Nitroimidazole

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-Nitro-1H-imidazole

ACTIAS31729G5EA 0 CHF
ACTIAS31729G25 ACTIAS31729G5
2-Nitroimidazole
2-Nitroimidazole
Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 279…284 °C
MDL Number: MFCD00005185
CAS Number: 527-73-1
UN: 2811
ADR: 6.1,III
Merck Index: 13,00923

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR