2-Nitrofluorene

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS34546G10EA 0 CHF
ACTIAS34546G10 ACTIAS34546G5 ACTIAS34546G100
2-Nitrofluorene
2-Nitrofluorene
Formula: C₁₃H₉NO₂
MW: 211.22 g/mol
Melting Pt: 154…157 °C
MDL Number: MFCD00001117
CAS Number: 607-57-8
EINECS: 210-138-5
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR