2-Nitro-p-xylene

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: Nitro-p-xylene2,5-Dimethylnitrobenzene

ACTIAS21370G100EA 0 CHF
ACTIAS21370G100
2-Nitro-p-xylene
2-Nitro-p-xylene
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1.132 g/cm³ (20 °C)
MDL Number: MFCD00024284
CAS Number: 89-58-7
EINECS: 201-920-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR