4-Aminocyclohexanol
Supplier: ACTIVATE SCIENTIFIC GMBH
ACTIAS32595G5EA
0
CHF
ACTIAS32595G5
ACTIAS32595G1
4-Aminocyclohexanol
4-Aminocyclohexanol
Formula:
C₆H₁₃NO |
CAS Number:
6850-65-3 EINECS: 229-943-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...