2-Amino-3-nitro-5-fluoropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS33525G5EA 0 CHF
ACTIAS33525G5 ACTIAS33525G1
2-Amino-3-nitro-5-fluoropyridine
2-Amino-3-nitro-5-fluoropyridine
Formula: C₅H₄FN₃O₂
MW: 157.1 g/mol
MDL Number: MFCD04972413
CAS Number: 212268-12-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR