2,3,4,5-Tetrafluorobenzoyl chloride

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: 2,3,4,5-tetrafluorobenzoylchloride

ACTIAS24955G100EA 0 CHF
ACTIAS24955G100
2,3,4,5-Tetrafluorobenzoyl chloride
2,3,4,5-Tetrafluorobenzoyl chloride
Formula: C₇HClF₄O
MW: 212.53 g/mol
Boiling Pt: 53…54 °C (1013 hPa)
Density: 1.58 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00075164
CAS Number: 94695-48-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR