Glycolic acid

Supplier: EHRENSTORFER GMBH
Danger

Synonyms: 2-Hydroxy acid2-Hydroxyacetic acid

EHERC14037500EA 74 CHF
EHERC14037500
Glycolic acid
Glycolic acid
Formula: C₂H₄O₃
MW: 76.05 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 79…80 °C
Density: 1.49 g/cm³ (20 °C)
Flash Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004312
CAS Number: 79-14-1
EINECS: 201-180-5
UN: 3261
ADR: 8,II
Merck Index: 13,04511

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR