DL-Tryptophan

Synonyms: (+/-)-2-Amino-3-(3-indolyl)propionic acid2-Amino-3-(1H-indol-3-yl)propanoic acidH-DL-Trp-OH

ACTIAS23861G100EA 0 CHF
ACTIAS23861G100
DL-Tryptophan
DL-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 289…290 °C
Density: 1.349 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064339
CAS Number: 54-12-6
EINECS: 200-194-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR