Indole-2-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 1H-Indole-2-carboxylic acid

ACTIAS7099G100EA 0 CHF
ACTIAS7099G100 ACTIAS7099KG1
Indole-2-carboxylic acid
Indole-2-carboxylic acid
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 203…206 °C
MDL Number: MFCD00005611
CAS Number: 1477-50-5
EINECS: 216-030-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR