2-Phenylpropionic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Danger

Synonyms: (±)-2-Phenylpropionic acid(±)-Hydratropic acidDL-2-Phenylpropionic acid(±)-2-Phenylpropanoic acid

ACTIAS21611G25EA 0 CHF
ACTIAS21611G25
2-Phenylpropionic acid
(±)-Hydratropic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: –5 °C
Density: 1,1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002650
CAS Number: 492-37-5
EINECS: 207-752-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR