6-Methoxy-1-indanone

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS7105G5EA 0 CHF
ACTIAS7105G25 ACTIAS7105G5 ACTIAS7105G1
6-Methoxy-1-indanone
6-Methoxy-1-indanone
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 108 °C
MDL Number: MFCD00021232
CAS Number: 13623-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR