2-Chlorothioanisole

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 2-Chlorophenyl methyl sulphide1-Chloro-2-(methylthio)benzene

ACTIAS20405G100EA 0 CHF
ACTIAS20405G25 ACTIAS20405G100
2-Chlorothioanisole
2-Chlorothioanisole
Formula: C₇H₇ClS
MW: 158.65 g/mol
MDL Number: MFCD00040124
CAS Number: 17733-22-1
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR