2,4-Dichlor-5-fluorpyrimidin

Lieferant: ACTIVATE SCIENTIFIC GMBH

ACTIAS50324G100EA 0 CHF
ACTIAS50324G100 ACTIAS50324G25 ACTIAS50324G5
2,4-Dichlor-5-fluorpyrimidin
2,4-Dichlor-5-fluorpyrimidin
Formel: C₄HCl₂FN₂
MW: 166,97 g/mol
MDL Number: MFCD00233551
CAS-Nummer: 2927-71-1
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR