2-Chloro-3-fluoropyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS50602G100EA 204 CHF
ACTIAS50602G5 ACTIAS50602G100
2-Chloro-3-fluoropyridine
2-Chloro-3-fluoropyridine
Formula: C₅H₃ClFN
MW: 131.54 g/mol
MDL Number: MFCD03095302
CAS Number: 17282-04-1
UN: 2810
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR