2-Chloronicotinic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

Synonyms: 2-Chloro-3-pyridinecarboxylic acid2-Chloropyridine-3-carboxylic acid

ACTIAS20138G250EA 0 CHF
ACTIAS20138G250 ACTIAS20138G50
2-Chloronicotinic acid
2-Chloronicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00006236
CAS Number: 2942-59-8
EINECS: 220-937-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR