3,6-Dichloropyridazine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS50578G100EA 0 CHF
ACTIAS50578G25 ACTIAS50578G500 ACTIAS50578G100
3,6-Dichloropyridazine
3,6-Dichloropyridazine
Formula: Câ‚„Hâ‚‚Clâ‚‚Nâ‚‚
MW: 148.98 g/mol
Boiling Pt: 89…91 °C (0.2 mmHg)
Melting Pt: 70 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006466
CAS Number: 141-30-0
EINECS: 205-478-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR