4-Chloroindole

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS51140G5EA 0 CHF
ACTIAS51140G25 ACTIAS51140G1 ACTIAS51140G5
4-Chloroindole
4-Chloroindole
Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 293 °C (1013 hPa)
Melting Pt: 84…86 °C
Density: 1,259 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005665
CAS Number: 25235-85-2
EINECS: 246-747-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR