2-Bromo-3-methoxypyridine

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS4048G1EA 0 CHF
ACTIAS4048G25 ACTIAS4048G1 ACTIAS4048G100
2-Bromo-3-methoxypyridine
2-Bromo-3-methoxypyridine
Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD01570896
CAS Number: 24100-18-3
EINECS: 246-017-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR