8-Bromoadenine
Supplier: Thermo Scientific
Warning
Synonyms: 8-Bromo-1H-adenine8-Bromo-9H-purin-6-amine
J65582.03EA 401 CHF
J65582.03 J65582.MC J65582.ME
8-Bromoadenine
8-Bromoadenine
Formula: C₅H₄BrN₅
MW: 214.02 g/mol
Melting Pt: >250 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00082518
CAS Number: 6974-78-3
EINECS: 230-225-1