2-Brom-3,5-difluorpyridin
Lieferant: ACTIVATE SCIENTIFIC GMBH
ACTIAS50652G5EA
0
CHF
ACTIAS50652G25
ACTIAS50652G5
ACTIAS50652G1
2-Brom-3,5-difluorpyridin
2-Bromo-3,5-difluoropyridine
Formel:
C₅H₂BrF₂N |
MDL Number:
MFCD03840197 CAS-Nummer: 660425-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...