(2-Bromophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 2-(2-Bromophenyl)acetic acid

ACTIAS40196G100EA 0 CHF
ACTIAS40196G100
(2-Bromophenyl)acetic acid
(2-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
MW: 215.05 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 107.5 °C
MDL Number: MFCD00004314
CAS Number: 18698-97-0
EINECS: 242-509-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR