2,4,6-Tribromopyrimidine

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS51645G1EA 0 CHF
ACTIAS51645G1 ACTIAS51645G5
2,4,6-Tribromopyrimidine
2,4,6-Tribromopyrimidine
Formula: C₄HBr₃N₂
MW: 316.78 g/mol
MDL Number: MFCD00233942
CAS Number: 36847-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR