1-Bromopyrene

Supplier: ACTIVATE SCIENTIFIC GMBH
Warning

ACTIAS24094G25EA 0 CHF
ACTIAS24094G25
1-Bromopyrene
1-Bromopyrene
Formula: C₁₆H₉Br
MW: 281.15 g/mol
MDL Number: MFCD00015767
CAS Number: 1714-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR