2,4,6-Trimethylphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC GMBH
Synonyms:
(2,4,6-Trimethylphenyl)metaborsäure
, Dihydroxy(2,4,6-trimethylphenyl)boran
, 2,4,6-Trimethylbenzolboronsäure
ACTIAS51618G5EA
0
CHF
ACTIAS51618G25
ACTIAS51618G1
ACTIAS51618G5
2,4,6-Trimethylphenylboronsäure
2,4,6-Trimethylphenylboronsäure
Formel:
C₉H₁₃BO₂ MW: 164,01 g/mol |
MDL Number:
MFCD00236060 CAS-Nummer: 5980-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...