3,4-Difluoro-DL-phenylalanine ≥97%
Supplier: Thermo Scientific
H32276.03EA 159 CHF
H32276.03 H32276.MD
3,4-Difluoro-DL-phenylalanine ≥97%
3,4-Difluoro-DL-phenylalanine
Formula: C₉H₉F₂NO₂
Melting Pt: 232…234 °C
Storage Temperature: Ambient
MDL Number: MFCD00061307
CAS Number: 32133-36-1