4'-(1-Pyrrolidinyl)acetophenone ≥98%
Supplier: Alfa Aesar
Warning
L15408.06EA 96.7 CHF
L15408.06
4'-(1-Pyrrolidinyl)acetophenone ≥98%
4'-(1-Pyrrolidinyl)acetophenone
Formula: C₁₂H₁₅NO
MW: 189.25
Melting Pt: 128…130 °C
Storage Temperature: Ambient
MDL Number: MFCD01075691
CAS Number: 21557-09-5
EINECS: 000-000-0