6-Methoxy-8-nitroquinoline

Supplier: ACTIVATE SCIENTIFIC GMBH

ACTIAS35069G10EA 0 CHF
ACTIAS35069G10 ACTIAS35069G5 ACTIAS35069G1
6-Methoxy-8-nitroquinoline
6-Methoxy-8-nitroquinoline
Formula: C₁₀H₈N₂O₃
MW: 204.1800 g/mol
MDL Number: MFCD00006802
CAS Number: 85-81-4
EINECS: 201-633-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR