4-Chloro-3-nitrophenylboronic acid

Supplier: ACTIVATE SCIENTIFIC GMBH

Synonyms: 4-Chloro-3-nitrobenzeneboronic acid

ACTIAS35155G1EA 0 CHF
ACTIAS35155G10 ACTIAS35155G1 ACTIAS35155G5
4-Chloro-3-nitrophenylboronic acid
4-Chloro-3-nitrophenylboronic acid
Formula: C₆H₅BClNO₄
MW: 201.37 g/mol
MDL Number: MFCD02258950
CAS Number: 151169-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR