2-Brombenzylamin Hydrochlorid

Lieferant: Molekula

MOLE36040140-5GEA 0 CHF
MOLE36040140-5G
2-Brombenzylamin Hydrochlorid
2-Brombenzylamin Hydrochlorid
Formel: C₇H₉BrClN
MW: 222,52 g/mol
MDL Number: MFCD00012853
CAS-Nummer: 5465-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR